• USA Home
  • MXSMCET24 - Milliflex Cassette prefilled with Cetrimide Agar with Naladixic Acid

EMAIL THIS PAGE TO A FRIEND
MXSMCET24 Millipore

Milliflex Cassette prefilled with Cetrimide Agar with Naladixic Acid

Recovery of Pseudomonas aeruginosa and inhibits growth of Klebsiella, Proteus, and Providencia species.

  •  eCl@ss 32160801

  •  NACRES NA.74

Purchase

Properties

Related Categories Microbiological Culture Media, Microbiology, Ready Media - Plates More...
Quality Level   300
form   plate (pre-poured agar)
shelf life   6 mo.
feature   ready-to-use
packaging   cassette of 10 mL
  pack of 24 tests
mfr. no.   Milliflex®
application(s)   bioburden testing: suitable
  microbiology: suitable
  water monitoring: suitable
color   light amberslightly opalescent
input   sample type cosmetics
sample type bottled water
sample type pharmaceutical(s)
sample type beer
pH   7.1±0.2 (25.0 °C)
Featured Industry   Cosmetics
Pharmaceutical
compatibility   for use with Milliflex®
shipped in   ambient
suitability   Pseudomonas aeruginosa
storage conditions   Right side up.
  2 to 8 °C

Description

General description

By combining the Milliflex products, accessories and services you can immediately streamline your microbial contamination testing workflow, leverage your quality assurance level by avoiding risks of exogenous contamination and ensure compliance to all GMP/GLP requirements.

Packaging

24 ea in 10 mL Agar in plastic Cassette

Application

Type of Test
Bioburden

Microbial Limits

Packaging

10 mL Agar in plastic Cassette

Specifications

Incubation Temperature: 37 °C

Incubation Time: 48 hrs

NF EN 12780 - ISO/CD16266 (project)

Culture Medium

Cetrimide

Analysis Note

P. aeruginosa species appear as green to blue colonies with fluorescence under UV wavelength

QC organisms
P. aeruginosa (ATCC 9027)

P. vulgaris (ATCC 13315) as negative control

Target Microorganisms: Pseudomonas aeruginosa

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?