• USA Home
  • MXSMCRA48 - Milliflex Cassette prefilled with R2A Agar

EMAIL THIS PAGE TO A FRIEND
MXSMCRA48 Millipore

Milliflex Cassette prefilled with R2A Agar

This low nutrient agar is used for the recovery of stressed heterotrophic plate count bacteria found in various types of waters, especially high purity water

  •  eCl@ss 32160801

  •  NACRES NA.74

Purchase

Properties

Related Categories Microbiological Culture Media, Microbiology, Ready Media - Plates More...
Quality Level   300
form   plate (pre-poured agar)
shelf life   12 mo.
feature   ready-to-use
packaging   cassette of 10 mL
  pack of 48 tests
mfr. no.   Milliflex®
application(s)   bioburden testing: suitable
  microbiology: suitable
  water monitoring: suitable
color   clear tan to light amber
input   sample type pharmaceutical(s)
sample type beer
sample type bottled water
sample type cosmetics
pH   7.2±0.2 (25.0 °C)
Featured Industry   Cosmetics
Pharmaceutical
compatibility   for use with Milliflex®
shipped in   ambient
suitability   heterotrophic bacteria (in a stressed environment)
storage conditions   Right side up.
  2 to 8 °C

Description

Packaging

48 ea in 10 mL Agar in plastic Cassette

Specifications

Incubation Temperature: 20–28 °C (standard method); 30–35 °C (EP)

Incubation Time: 120–168 hrs (standard method); 120 hrs (EP)

Standard Methods for the Examination of Water and Wastewater, European Pharmacopeia 2002, addendum 4.6

Culture Medium

R2A

Analysis Note

Target Microorganisms: Heterotrophic bacteria in a stressed environment

Safety & Documentation

Safety Information

WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?