• USA Home
  • MXSMCTS48 - Milliflex Cassette prefilled with Tryptic Soy Agar

EMAIL THIS PAGE TO A FRIEND
MXSMCTS48 Millipore

Milliflex Cassette prefilled with Tryptic Soy Agar

This medium is used for the recovery of a broad range of heterotrophic microorganisms such as common aerobic and facultative anaerobic bacteria found in various types of water

  •  eCl@ss 32160801

  •  NACRES NA.74

Purchase

Properties

Related Categories Microbiological Culture Media, Microbiology, Ready Media - Plates More...
Quality Level   300
form   plate (pre-poured agar)
shelf life   12 mo.
feature   ready-to-use
packaging   cassette of 10 mL
  pack of 48 tests
mfr. no.   Milliflex®
application(s)   bioburden testing: suitable
  microbiology: suitable
  water monitoring: suitable
color   tan to light amber
input   sample type bottled water
sample type cosmetics
sample type beer
sample type pharmaceutical(s)
pH   7.3±0.2 (25.0 °C)
Featured Industry   Cosmetics
Pharmaceutical
compatibility   for use with Milliflex®
shipped in   ambient
suitability   heterotrophic bacteria
  heterotrophic bacteria
storage conditions   Right side up.
  2 to 8 °C

Description

General description

By combining the Milliflex products, accessories and services you can immediately streamline your microbial contamination testing workflow, leverage your quality assurance level by avoiding risks of exogenous contamination and ensure compliance to all GMP/GLP requirements.

Packaging

48 ea in 10 mL Agar in plastic Cassette

Application

Type of Test
Bioburden

Microbial Limits

Packaging

10 mL Agar in plastic Cassette

Specifications

Incubation Temperature: 30–35 °C

Incubation Time: 48–72 hrs (USP); 120 hrs max (EP)

United-States Pharmacopeia, European Pharmacopeia

Culture Medium

Trypticase Soy

Analysis Note

Colonies appear clear to creamy white, some may produce pigment

QC organisms
E. coli (ATCC 25922)

P. aeruginosa (ATCC 9027)

S. aureus (ATCC 6538)

B. subtilis (ATCC 6633)

C. albicans (ATCC 10231)

A. niger (ATCC 16404)

Target Microorganisms: Heterotrophic microorganisms

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?