• USA Home
  • MXSMHPC48 - Milliflex Cassette prefilled with Heterotrophic Plate Count Agar

EMAIL THIS PAGE TO A FRIEND
MXSMHPC48 Millipore

Milliflex Cassette prefilled with Heterotrophic Plate Count Agar

This medium is used for the recovery of stressed heterotrophic plate count bacteria found in various types of waters, especially high purity water

  •  eCl@ss 32160801

  •  NACRES NA.74

Purchase

Properties

Related Categories Microbiological Culture Media, Microbiology, Ready Media - Plates More...
Quality Level   300
form   plate (pre-poured agar)
shelf life   6 mo.
feature   ready-to-use
packaging   cassette of 10 mL
  pack of 48 tests
mfr. no.   Milliflex®
application(s)   bioburden testing: suitable
  microbiology: suitable
  water monitoring: suitable
color   clear tan to light amber
input   sample type beer
sample type cosmetics
sample type bottled water
sample type pharmaceutical(s)
pH   7.1±0.2 (25.0 °C)
Featured Industry   Cosmetics
Pharmaceutical
compatibility   for use with Milliflex®
shipped in   ambient
storage temp.   2-8°C
suitability   heterotrophic bacteria
  heterotrophic bacteria
storage conditions   Right side up.
  2 to 8 °C

Description

General description

Milliflex® Cassette are ready to use agar-based media to be used with Milliflex® or Milliflex Oasis® system for convenient, rapid and reliable bioburden testing. The prefilled agar cassette helps to streamline the microbial contamination testing workflow with quality assurance and avoiding risks of exogenous contamination. Each plate is a 3-piece assembly containing approximately 10ml of the media. Heterotrophic Plate Count, formerly known as the standard plate, is a procedure for estimating the number of live heterotrophic bacteria in potable water. Peptic digest of animal tissue is the source of nutrients for organisms, which are not highly fastidious. Gelatin is utilized by microorganisms through a proteolytic mechanism and the addition of glycerol to the basal medium provides a source of carbon and energy.

Application

Milliflex® Cassette prefilled with Heterotrophic Plate Count Agar is used in Bioburden testing and general microbiological analysis of water especially high purity water (electronics, RO, distilled or purified). It is widely used in pharmaceutical, cosmetic and brewing industry.

Packaging

48 ea in 10 mL Agar in plastic Cassette

Culture Medium

Heterotrophic Plate Count

Analysis Note

Colonies appear clear to creamy white, some may produce pigment

QC organisms
E. coli (ATCC 25922)

P. aeruginosa (ATCC 9027)

S. aureus (ATCC 6538)

B. subtilis (ATCC 6633)

Target Microorganisms: Heterotrophic microorganisms

Legal Information

MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Milliflex® and Milliflex Oasis® Solid Culture Media and Cassettes

We offer a full range of ready-to-use agar media cassettes manufactured according to ISO certified processes with high purity ingredients for long life to use with the Milliflex Oasis® system for bio...
Keywords: Aerobic, Anaerobic

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?