• USA Home
  • NXF-01 - NatriFlo HD-Q Recon Mini

EMAIL THIS PAGE TO A FRIEND
NXF-01 Millipore

NatriFlo HD-Q Recon Mini

  •  eCl@ss 32011308

Purchase

Properties

Related Categories Biopharmaceutical Manufacturing, Biopharmaceutical Process Development and Manufacturing, Chromatography for Biopharmaceutical Manufacturing More...
mfr. no.   NatriFlo
parameter   1-5 mL/min flow rate
bed H   0.5 mm
bed volume   0.2 mL
pore size   0.4 μm nominal pore size
capacity   0.04 g binding capacity (BSA)(per device)
shipped in   ambient

Description

General description

With a revolutionary three-dimensional macroporous hydrogel structure that provides a high density of binding sites and rapid mass transfer, Natrix® HD membranes deliver binding capacity that exceeds resin-based columns with fast flow rates. This combination of performance and speed enables low-risk, scalable solutions for efficient purification of biologics and eliminates costly packing, cleaning, validating and other burdens of conventional methods. The single-use membrane chromatography technology can be utilized for initial capture steps all the way through polishing and is adaptable to most ligand chemistries or affinity products.
NatriFlo HD-Q Chromatography Membrane Adsorber is a high capacity, high throughput strong anion exchange membrane adsorber for single-use per batch biomolecule purification applications.
The NatriFlo HD-Q Recon Mini is a scaled down, single use chromatography laboratory model to screen and fine-tune buffer conditions and load parameters before moving to small-scale and commercial manufacturing.

mAb nominal polishing capacity: 2 g

Membrane configuration: Flat Sheet
10 devices per package

Legal Information

NATRIFLO is a trademark of Natrix Separations Inc.

NATRIX is a registered trademark of Natrix Separations Inc.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Comparison of process-related impurity clearance for membrane adsorbers and resin-based chromatography technologies

Strong anion exchange (Q) chromatography has become an industry standard in polishing steps for monoclonal antibody (mAb) purification. It is a proven technology to remove DNA, viruses, endotoxins an...
Keywords: Buffers, Cell culture, Chromatography, Column chromatography, Ligands, Mass spectrometry, Polymerase chain reaction - quantitative, Purification, Reductions, Separation

Robust, load independent viral clearance in monoclonal antibody purification with NatriFlo® HD-Q membrane adsorber

Anion exchange chromatography is an important flowthrough polishing step for viral clearance of monoclonal antibodies (mAbs). Resin based materials are most commonly used for this but the large size ...
Keywords: Buffers, Cell culture, Chromatography, Clinical, Column chromatography, Diffusion, Ligands, Mass spectrometry, Reductions

Related Content

Coronavirus COVID-19 (SARS-CoV-2) Detection, Characterization & Vaccine Production

As a global life science tools and equipment supplier, we are here to help in the response to COVID-19. From supplying raw materials and research products for detection and characterization to produc...
Keywords: PAGE

Natrix® Membrane Chromatography Technology

The NatriFlo® HD-Q membrane is a porous polyacrylamide hydrogel containing a high density of pendant quaternary ammonium (Q) binding groups physically reinforced by an inert fiber web. The interconne...
Keywords: Chromatography, Ligands, Purification, Reductions, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?