EMAIL THIS PAGE TO A FRIEND
OX0130 Supelco

1-Octanol

Synonym: 1-Octanol, n-Octyl Alcohol, N-Octanol

  • CAS Number 111-87-5

  • Linear Formula CH3(CH2)6CH2OH

  •  MDL number MFCD00002988

  •  EC Index Number 203-917-6

Purchase

Properties

Quality Level   200
vapor pressure   0.031 hPa ( 20 °C)
assay   ≥98% (GC)
form   liquid
bp   195 °C/1013 hPa
transition temp   flash point 86 °C
solubility   0.43 g/L (External MSDS)
density   0.83 g/cm3 at 20 °C
shipped in   ambient
InChI   1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3
InChI key   KBPLFHHGFOOTCA-UHFFFAOYSA-N
storage conditions   Store at room temperature.

Description

Packaging

4 l in Glas bottle pre packed

500 ml in Glas bottle pre packed

Analysis Note

Assay (GC): 98% min
Color (APHA): 10 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
WGK Germany 
WGK 1
Flash Point(F) 
187.7 °F
Flash Point(C) 
86.5 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?