• USA Home
  • PK1111 - Anti-GSK-3β Rabbit pAb

EMAIL THIS PAGE TO A FRIEND
PK1111 Sigma-Aldrich

Anti-GSK-3β Rabbit pAb

liquid, clone, Calbiochem®

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, GQ-GZ, Primary Antibodies
clone   polyclonal
biological source   rabbit
species reactivity   rat, human, mouse
form   liquid
shipped in   wet ice
isotype   IgG
Quality Level   100
antibody product type   primary antibodies
contains   0.01% thimerosal as preservative
mfr. no.   Calbiochem®
storage condition   OK to freeze
  avoid repeated freeze/thaw cycles
storage conditions   -20C

Description

General description

Anti-GSK-3β, rabbit polyclonal, recognizes the ~46 kDa GSK-3β protein in mouse small intestine tissue. It is validated for use in Western blotting and immunohistochemistry.

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~46 kDa GSK-3 protein.

Recognizes the ~46 kDa GSK-3β protein in mouse small intestine tissue.

Immunogen

Human

recombinant human GSK-3β

Application


Immunoblotting (0.5-4 µg/ml)

Immunohistochemistry (10-30 µg/ml; see comments)

Physical form

In PBS, 30% glycerol, 0.5% BSA.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C) for long-term storage.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Analysis Note

Positive Control
Mouse small intestine

Other Notes

Garrido, J.J., et al. 2007.FEBS Lett.8, 1579.
Meares, G.P. and Jope, R.S. 2007.J Biochem.23, 16989.

This antibody has been reported to work for immunohistochemistry in the range of 10-30 µg/ml, but the optimal concentration should be determined for each experiment. Antibody should be titrated for optimal results in individual systems.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?