• USA Home
  • PX1550 - Potassium Permanganate

EMAIL THIS PAGE TO A FRIEND
PX1550 Supelco

Potassium Permanganate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym: Permanganic Acid Potassium Salt

  • CAS Number 7722-64-7

  • Empirical Formula (Hill Notation) KMnO4

  • Molecular Weight 158.03

  •  Colour Index Number 77755

  •  EC Index Number 025-002-00-9

Purchase

Properties

packaging   2.5 kg in Plastic bottle (PX1550-5)
  50 kg in Drum, fibre (PX1550-20)
mp   >240 °C (decomposition)
pH   7-9 (20 °C, 20 g/L in H2O)
shipped in   ambient
vapor pressure   <0.01 hPa ( 20 °C)
solubility   64 g/l
storage conditions   Store at room temperature.

Description

Analysis Note

Appearance: Dark solid
Assay (iodometric): 99.0% min
Chloride and chlorate (as Cl): 0.005% max
Insoluble matter: 0.2% max
Sulfate (SO4): 0.02% max

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Packaging

2.5 kg in Plastic bottle

50 kg in Drum, fibre

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?