EMAIL THIS PAGE TO A FRIEND
PX1827 Supelco

Isopropyl Alcohol

anhydrous, septum-sealed bottle DriSolv®

Synonym: 2-Propanol,IPA, 2-Propanol

Purchase

Properties

packaging   100 ml in Glass bottle (PX1827-7)
bp   82.4 °C (1013 hPa)
vapor pressure   43 hPa (20 °C)
flash point   12 °C
mp   -89.5 °C
shipped in   ambient
storage temp.   Store at room temperature.
specifications   Appearance: Clear liquid, free from particulates
  Assay (GC): 99.9% min
  Color (APHA): 10 max
  Evaporation residue: 1ppm max
  Filtered through 0.2 µm filter: To pass test
  Water (H2O): 50ppm max
Safety & Documentation

Safety Information

RTECS 
NT8050000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly hazardous to water
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Irritant  Irritant
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?