EMAIL THIS PAGE TO A FRIEND
R04587 Sigma-Aldrich

Formalin

ready-to-use solution, HARLECO®, 20%

Synonym: NBF Fixative, Formalin 10-20%

Purchase

Properties

Quality Level   300
form   ready-to-use solution
mfr. no.   HARLECO®
concentration   20%
application(s)   hematology: suitable
  histology: suitable
color   colorless
Featured Industry   Diagnostic Assay Manufacturing
shipped in   ambient
storage temp.   room temp
storage conditions   Store at +15°C to +30°C.

Description

General description

HARLECO® Ready-to-use fixing solution, 20% Neutral Buffered Formalin (NBF)

Packaging

1 Case in Plastic bottle

Application

HARLECO® Ready-to-use fixing solution, 20% Neutral Buffered Formalin (NBF)

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Target organs 
Respiratory system
WGK Germany 
WGK 3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?