• USA Home
  • SC00B10RE - Stericup Receiver Flask

EMAIL THIS PAGE TO A FRIEND
SC00B10RE Millipore

Stericup Receiver Flask

1000 mL, radio-sterilized, 12 flasks, for the storage of sterilized or clarified fluids

  •  eCl@ss 32031623

Purchase

Properties

packaging   12 pack, Receiver Flask, 1000 mL, radio-sterilized (SC00B10RE)
H   183 mm
compatibility   for use with Steritop Filter Units
diam.   45 mm
fitting   inlet thread(s) 45 mm
  outlet bottle top (Universal)
material   polystyrene
mfr. no.   Stericup
parameter   1000 mL process volume
  45 °C max. temp.
shipped in   ambient
test parameters   sample volume: 1 L
sterility   sterile
hold-up volume   Not Applicable

Description

Application

1000 mL, radio-sterilized, 12 flasks, for the storage of sterilized or clarified fluids

General description

1000 mL bottle with cap

Steritop bottle top filter units provide a large filter area for rapid sterile filtration of aqueous fluids into narrow-mouth (33 mm) or wide-mouth (45 mm) bottles. A small tab inside the funnel secures an optional prefilter in-place during filtration. For added protection against contamination, the threads for bottle attachment have been slightly recessed in case the funnel accidentally touches a work surface.

Preparation Note

Sterilization Method
Gamma irradiation

Physical form

Color Code: Clear with blue cap

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Stem Cell Testing of PES Membrane Containing Stericup® Quick Release Filters

Though microporous filtration is the method of choice to prepare and minimize contamination of cell culture media and reagents, there still remains some concern that the filtration step will remove v...
Keywords: Buffers, Capture ELISA, Cell culture, Culture media, Enzyme-linked immunosorbent assay, Filtration, Gene expression, Growth factors, Immunocytochemistry, Sterilizations, Transcription

Related Content

Filter Membrane Selection Recommendations

What are you filtering out? What do you want to keep? Pick the right membrane for your filtration.
Keywords: Cell culture, Filtration

Stericup® and Steritop® Filtration Systems

Stericup® Sterile Vacuum Filtration Systems are known to research scientists worldwide for reliable, high-performance membranes and optimum sterile filtration of laboratory reagents, buffers, and med...
Keywords: Buffers, Culture media, Filtration, Sterilizations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?