• USA Home
  • ST1631 - Anti-BMP7 Mouse mAb (M1-F8)

EMAIL THIS PAGE TO A FRIEND
ST1631 Sigma-Aldrich

Anti-BMP7 Mouse mAb (M1-F8)

liquid, clone M1-F8, Calbiochem®

Synonym: Anti-Bone Morphogenetic Protein 7

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, BL-BZ, Primary Antibodies
clone   M1-F8, monoclonal
biological source   mouse
species reactivity   mouse, human
form   liquid
shipped in   wet ice
isotype   IgG1
Quality Level   100
antibody product type   primary antibodies
does not contain   preservative
mfr. no.   Calbiochem®
storage condition   OK to freeze
  avoid repeated freeze/thaw cycles
storage conditions   -20C

Description

General description

Anti-BMP7, mouse monoclonal, clone M1-F8, recognizes the ~45-50 kDa BMP7 protein in A-549 and Raw 264.7 cells and in human uterine myoma. It is validated for Western blotting.

Purified mouse monoclonal antibody. Recognizes the ~45-50 kDa BMP7 protein.

Recognizes the ~45-50 kDa BMP7 protein in A-549 and Raw 264.7 cells and human uterine myoma.

Immunogen

Full-length, recombinant, human BMP7 (aa 1-432), expressed as a GST fusion protein

Application




Immunoblotting (1-5 µg/ml)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Warning

Toxicity: Regulatory Review (Z)

Analysis Note

Negative Control
293T cells

Positive Control
Human uterus myoma, A-549 cells, Raw 264.7 cells

Other Notes

Loureiro, J., et al. 2010. Nephrol. Dial Transplant25 1098.

Variables associated with assay conditions will dictate the proper working dilution.

Safety & Documentation

Safety Information

WGK Germany 
nwg
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?