EMAIL THIS PAGE TO A FRIEND
SX0210 Supelco

Silver Sulfate

Meets ACS Specifications GR ACS

Synonym: Silver Sulfate

  • CAS Number 10294-26-5

  • Empirical Formula (Hill Notation) Ag2O4S

  • Molecular Weight 311.80

  •  MDL number MFCD00003407

  •  EC Index Number 233-653-7

Purchase

Properties

Quality Level   200
pH   5-6 (25 °C, 5 g/L in H2O)
mp   655 °C
solubility   8 g/L
shipped in   ambient
InChI   1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;
InChI key   YPNVIBVEFVRZPJ-UHFFFAOYSA-N
storage conditions   Store at room temperature.

Description

Packaging

25, 100 g in Plastic bottle

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay: 98.0% min
Color: White
Form: Fine crystals
Insoluble matter and silver chloride: 0.02% max
Iron (Fe): 0.001% max
Nitrate (NO3): 0.001% max
Substances not precipitated by HCl: 0.03% max

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?