• USA Home
  • SX1243M - Sulfuric acid solution (6.0N)

EMAIL THIS PAGE TO A FRIEND
SX1243M Supelco

Sulfuric acid solution (6.0N)

Synonym: Sulfuric acid solution, Sulphuric Acid

  • CAS Number 7664-93-9

  • Linear Formula H2SO4

  • Molecular Weight 98.08

  •  Beilstein/REAXYS Number 2037554

  •  MDL number MFCD00064589

Purchase

Properties

Related Categories Acids and Bases, Biochemicals and Reagents, Denaturation More...
Quality Level   100
bp   103 °C/1013 hPa
Agency/Method   traceable to NIST
shipped in   ambient
SMILES string   OS(O)(=O)=O
InChI   1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)
InChI key   QAOWNCQODCNURD-UHFFFAOYSA-N
storage conditions   No declaration

Description

Packaging

1 l in Plastic bottle

Specifications

Tolerance: ±0.030N

Analysis Note

Color Limit: "10 max."

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?