• USA Home
  • XX1004732 - Glass base with screen, 47 mm

EMAIL THIS PAGE TO A FRIEND
XX1004732 Millipore

Glass base with screen, 47 mm

Accessories for filter holders for sample preparation.

Purchase

Properties

Related Categories Filtration Supplies, Labware, Membrane Holders and Assemblies More...
fitting outlet   No. 8 perforated silicone stopper mounts in standard 1 L and 4 L filtering flasks
diameter   47 mm
filtration area   9.6 cm²
material   Borosilicate glass base, stainless steel screen
material size   1
shipped in   ambient

Description

Background information

REMOVE FROM SCOPE

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?