• USA Home
  • XX1009000 - Funnel, 1L, 90 mm, ground glass seal

EMAIL THIS PAGE TO A FRIEND
XX1009000 Millipore

Funnel, 1L, 90 mm, ground glass seal

Accessories for filter holders for sample preparation.

Purchase

Properties

Related Categories Filtration Supplies, Labware, Membrane Holders and Assemblies More...
diameter   90 mm
flask capacity   1 L
material   Borosilicate glass
material size   1
shipped in   ambient

Description

Background information

Accessories for Glass Filter Holders – XX100900, XX1604705, XX1604706, XX1009010, XX1009002


The All-Glass Filter Holder was designed for the filtration of aqueous and organic or corrosive liquids for particulate contamination analysis. The holder can also be used for HPLC solvent preparation. The vacuum connection is integrated into the filter holder base and flask cap, above the filtrate exit level. With this design, you can avoid unintentionally drawing filtrate into vacuum tubing. This design also allows simple transfer of filtrate (i.e., HPLC solvents) by pouring from the receiver flask.

These part numbers are accessory items for the all glass filter holders.

Features & Benefits:
•All-glass construction has broad chemical compatibility
•Borosilicate glass parts contact liquid, with ground-glass sealing surfaces
•Vacuum connection to flask cap simplifies transfer of filtrate

Applications:
Particle Contamination Analysis, HPLC Solvent Filtration, General Filtration and Clarification

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?