• USA Home
  • XX1104706 - Cover for Sterifil receiver flask

EMAIL THIS PAGE TO A FRIEND
XX1104706 Millipore

Cover for Sterifil receiver flask

Purchase

Properties

Related Categories Filtration Supplies, Labware, Membrane Holders and Assemblies More...
trade name   Sterifil
sterility   Sterile
diameter   47 mm
volume   Not applicable
material   Polysulfone receiver flask cover
device material   Polysulfone (PSF)
material size   1
shipped in   ambient

Description

Background information

The Sterifil® aseptic system was designed as a tool for general filtration applications and filtration of samples for particulate or biological contamination analysis. The closed unit protects sample and filtrate from environmental contamination. Sterifil® holder and funnel are available separately (without receiver flask and cover) to use with standard 1 L filter flasks or multiple place manifolds.

Depending upon membrane filter selection, the Sterifil® products can be used for a variety of applications including bioburden monitoring and enumeration of organisms in samples.

Features & Benefits:
•Closed unit protects samples and filtrate from environmental contamination
•Device is autoclavable with membrane in place for sterile applications with less waste
•Optional hand pump allows for on-site filtration

Applications:
General Filtration, Field-based Filtration (with optional hand pump), Bioburden Monitoring, Counting Microorganisms, Filtration Processes Requiring Membrane to be Retained

applicable device

Sterifil Aseptic System

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Articles

Sterifil® Aseptic System and Filter Holder

Sterifil® devices are ideal for vacuum filtration of liquids for analyzing microbiological or particulate content. Choose from two configurations, the closed Sterifil® aseptic filtration system and t...
Keywords: Filtration

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?