• USA Home
  • XX1504702 - Glass Base & Cap, 47 mm

EMAIL THIS PAGE TO A FRIEND
XX1504702 Millipore

Glass Base & Cap, 47 mm

Accessories for filter holders for sample preparation.

Purchase

Properties

Related Categories Filtration Supplies, Labware, Membrane Holders and Assemblies More...
fitting outlet   6 mm (1/4 in.) O.D. tubulated cap sidearm to vacuum
diameter   47 mm
filtration area   9.6 cm²
material   Borosilicate glass base and tubulated cap
material size   1
shipped in   ambient

Description

Background information

REMOVE FROM SCOPE

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?