• USA Home
  • XX2004702 - Base with screen, gasket and stopper

EMAIL THIS PAGE TO A FRIEND
XX2004702 Millipore

Base with screen, gasket and stopper

Accessories for filter holders for sample preparation.

Purchase

Properties

Related Categories Filtration Supplies, Labware, Membrane Holders and Assemblies More...
fitting outlet   No. 8 perforated silicone stopper mounts in standard 1 L filtering flask
diameter   47 mm
filtration area   9.6 cm²
material   Stainless steel base and filter screen, silicone stopper, PTFE gasket
material size   1
shipped in   ambient

Description

Background information

The Hydrosol filter holder is designed for the filtration of flammable liquids. Unlike other filter holders, the Hydrosol filter holder includes a grounding unit to prevent sparking from static discharge, which can be a safety hazard.

Applications:
Vacuum Filtration of Flammable Liquids

XX2004720 Hydrosol Filter Holder, 47 mm, stainless steel

Replacement Parts
XX2004704 Funnel and Locking Ring Assembly, stainless steel
XX2004708 Support Screen, 47 mm, stainless steel
XX2004702 Base with Screen, gasket and support
XX2004703 Gasket, PTFE
XX2004718 No. 8 Stopper, 9.5 mm (3/8 in.) hole, silicone
XX20047RK Replacement Parts Kit for Hydrosol Filter Holder

Accessories
XX1004705 Vacuum Filtering Flask, 1 L
XX1004744 Vacuum Filtering Flask, 4L
XX7100004 Tubing 3/16 in. I.D. x 140 cm, silicone
XX6200006P Filter Forceps, blunt end, stainless steel
XX2604735 3-place Manifold, 47 mm, PVC
XX2504735 3-place Manifold, 47 mm, stainless steel
XX2504700 6-place Manifold, 47 mm, stainless steel
WP6211560 High Output Pump, 115 V/60 Hz
WP6222050 High Output Pump, 220 V/50 Hz
WP6210060 High Output Pump, 100 V/50-60 Hz

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?