• USA Home
  • XX2504738 - 3-Place Sterility Test Manifold

EMAIL THIS PAGE TO A FRIEND
XX2504738 Millipore

3-Place Sterility Test Manifold

  •  eCl@ss 32014001

  •  NACRES NB.24

Purchase

Properties

Related Categories Labware, Manufacturer Browser, Millipore - Manufacturer Browser, Millipore Filters, Millipore Filtration Vacuum,
application(s)   bioburden testing: suitable
  sterility testing: suitable
input   sample type cosmetics
sample type pharmaceutical(s)
sample type bottled water
sample type beer
Featured Industry   Cosmetics
Environmental
Food and Beverages
shipped in   ambient

Description

General description

Stainless steel manifolds for sterility testing

Application

Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Bioburden Testing, Water Monitoring

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?