• USA Home
  • XX3002561 - 1/8 in. NPTM to M Luer locking nickel/chrome plated
EMAIL THIS PAGE TO A FRIEND
XX3002561 Millipore
1/8 in. NPTM to M Luer locking nickel/chrome plated
Accessories for filter holders for sample preparation.
Purchase
Properties
Related Categories Filtration Supplies, Labware, Membrane Holders and Assemblies More...
diameter   0.3 cm (0.125 in.) (I.D.)
material   Nickel/Chrome plated
material size   1
shipped in   ambient
Description
Background information
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.
Tri-Clamp (TC) fittings are referred to by convention as 1 1/2", 2", etc. 1 1/2" TC fittings connect to 1" or 1 1/2" O.D. tube sizes. 2" TC fittings connect to 2" O.D. tube size only.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?