EMAIL THIS PAGE TO A FRIEND
XX4004716 Millipore

O-rings, PTFE

Accessories for filter holders for sample preparation.

Purchase

Properties

Related Categories Filtration Supplies, Labware, Membrane Holders and Assemblies More...
material size   5
shipped in   ambient

Description

Background information

These filter holders are ideal for ultracleaning or sterilizing batch volumes of liquids. The cylindrical barrel holds 100 mL (or 340 mL) of liquid, which is filtered by externally applied gas pressure through a suitable filter supported on the holder base.

Features & Benefits:
•Filter holder has reservoir to hold 100 mL (or 340 mL) of liquid for batch filtration
•Autoclavable with membrane in place for sterile filtration

Applications:
Batch Filtration

applicable device

Stainless Steel Filter Holder

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?