• USA Home
  • 10106801001 - Kanamycin sulfate

EMAIL THIS PAGE TO A FRIEND
10106801001 Roche

Kanamycin sulfate

Purchase

Properties

Related Categories Antibiotics, Core Bioreagents, Research Essentials More...
description   C18H36N4O11 x H2SO4 (Formula)
assay   (Chromatographically homogeneous)
form   powder
mol wt   Mr 485.5 (Kanamycin)
  Mr 582.6 (Kanamycin sulfate)
packaging   pkg of 5 g
mfr. no.   Roche
shipped in   wet ice
storage temp.   2-8°C

Description

Biochem/physiol Actions

Kanamycin sulfate is an aminoglycosidic antibiotic with broad-spectrum inhibition of the growth of Gram-positive and Gram-negative bacteria and mycoplasmas in cell culture. It is also used to study cell-free protein biosynthesis.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibiotic Kill Curve

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic to determine the minimum concentration of an antibiotic that c...
Keywords: Antibiotics, Cell culture, Evaporation, Transfection

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?