• USA Home
  • 11117831001 - cDNA Synthesis Kit

EMAIL THIS PAGE TO A FRIEND
11117831001 Roche

cDNA Synthesis Kit

Purchase

Properties

Quality Level   100
mfr. no.   Roche
packaging   kit of for up to 10 reactions

Description

General description

cDNA Synthesis System; Instructions For Use

Optimized one-tube procedure for the synthesis of double-stranded cDNA up to 3 kb from total RNA or mRNA.

The cDNA Synthesis System uses an optimized one-tube procedure, based on the method of Gubler and Hoffmann.
Sample: 1 - 20 μg total RNA, 0.3 - 2 μg mRNA
Assay time: 60 minutes for 1st strand synthesis, 120 minutes for 2nd strand synthesis
Product: double-stranded cDNA copy of an RNA sample

Application

Optimized one-tube procedure for the synthesis of double-stranded cDNA from total RNA or mRNA.
The ds cDNA can be used for the construction of non-directional cDNA libraries, as a starting point for substractive hybridization experiments to enrich differentially expressed genes, and for the in vitro transcription of whole cDNA populations to generate labeled cRNA for hybridization on DNA microarrays.

Components

• AMV Reverse Transcriptase Buffer, 5x concentrated
• AMV Reverse Transcriptase (25 U/μl)
• 1,4-Dithiothreithol (0.1 M)
• Protector RNase Inhibitor (25 U/μl)
• Oligo (dT)15 primer, 200 μM (1 μg/μl)
• Oligo [(dT24 )T7promotor]65 primer, 100 μM (2 μg/μl)
• dNTP mixture (each nucleotide, 10 mM)
• Control neo mRNA (0.2 μg/μl)
• 2nd strand synthesis buffer, 5x concentrated
• 2nd strand enzyme blend (mixture of DNA Polymerase I, E. coli Ligase and RNase H)
• T4 DNA Polymerase (1 U/μl)
• Double-distilled water
• RNase I (10 U/μl)
• Proteinase K, recombinant, PCR grade (50 U/mll)

Other Notes

Reverse Transcriptases Overview

For life science research only. Not for use in diagnostic procedures.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?