EMAIL THIS PAGE TO A FRIEND
TRNAMRE-RO Roche

tRNA

from E. coli MRE 600

Purchase

Properties

Quality Level   100
form   lyophilized
packaging   pkg of 100 mg (10109541001)
  pkg of 500 mg (10109550001)
mfr. no.   Roche
shipped in   dry ice
storage temp.   −20°C

Description

General description

Transfer RNAs (tRNA) are responsible for amino acid transport and the correct reading frame in protein biosynthesis. The knowledge of their tertiary structure and interactions with proteins and ribosomes is important for the understanding of mechanisms involved in protein biosynthesis.

Application

tRNA is added as a competitor to pre- and hybridization solutions to prevent nonspecific binding of the probe.

Quality

This product is tested for the absence of RNase according to the current Quality Control procedures. 1mg of dry substance corresponds to 16A260 units.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?