• USA Home
  • 03-0066 - Barbital buffer solution

EMAIL THIS PAGE TO A FRIEND
03-0066 Sigma-Aldrich

Barbital buffer solution

pH 8.6, 10 mM, electrophoretic mobility (μ) 0.06

Synonym: Buffer concentrate from sodium barbital and barbital, Sodium barbital buffer solution

Purchase

Properties

drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
availability   available only in Japan
concentration   10 mM
electrophoretic mobility (μ)   0.06
pH   8.6

Description

Suitability

for electrophoresis

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?