EMAIL THIS PAGE TO A FRIEND
03-0120 Sigma-Aldrich

Barium acetate

JIS special grade, ≥99.0%

Purchase

Properties

grade   JIS special grade
assay   ≥99.0%
availability   available only in Japan
SMILES string   CC(=O)O[Ba]OC(C)=O
InChI   1S/2C2H4O2.Ba/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2
InChI key   ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
AF4550000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?