• USA Home
  • 03-3070 - Boron trifluoride-methanol solution

EMAIL THIS PAGE TO A FRIEND
03-3070 SAJ

Boron trifluoride-methanol solution

CP

  • CAS Number 373-57-9

  • Linear Formula BF3 · MeOH

  • Molecular Weight 99.85

  •  Beilstein Registry Number 3611499

  •  MDL number MFCD00071635

Purchase

Properties

grade   CP
InChI Key   JBXYCUKPDAAYAS-UHFFFAOYSA-N
availability   available only in Japan
concentration   ~68% boron trifluoride
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?