EMAIL THIS PAGE TO A FRIEND
05-0075 Sigma-Aldrich

Cadmium

≥99.9%

  • CAS Number 7440-43-9

  • Empirical Formula (Hill Notation) Cd

  • Molecular Weight 112.41

  •  Beilstein/REAXYS Number 8137359

  •  EC Number 231-152-8

  •  MDL number MFCD00010914

  •  PubChem Substance ID 329748003

Purchase

Properties

Related Categories Cadmium Catalysts, Catalysis and Inorganic Chemistry, Chemical Synthesis More...
assay   ≥99.9%
reaction suitability   reagent type: catalyst
core: cadmium
availability   available only in Japan
resistivity   7.27 μΩ-cm, 22°C
particle size   0.5-2 mm
bp   765 °C (lit.)
mp   320.9 °C (lit.)
density   8.65 g/mL at 25 °C (lit.)
SMILES string   [Cd]
InChI   1S/Cd
InChI key   BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Description

Suitability

for determination of nitrate

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN3288 - class 6.1 - PG 2 - EHS - Toxic solid, inorganic, n.o.s., HI: all
WGK Germany 
2
RTECS 
EU9800000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?