EMAIL THIS PAGE TO A FRIEND
05-6560 Sigma-Aldrich

m-Cresol Purple

JIS special grade

Synonym: m-Cresolsulfonphthalein

  • CAS Number 2303-01-7

  • Empirical Formula (Hill Notation) C21H18O5S

  • Molecular Weight 382.43

  •  Beilstein Registry Number 350314

  •  EC Number 218-960-6

  •  MDL number MFCD00005871

  •  PubChem Substance ID 329748139

Purchase

Properties

Related Categories C, Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
grade   JIS special grade
InChI Key   OLQIKGSZDTXODA-UHFFFAOYSA-N
availability   available only in Japan
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?