• USA Home
  • 09-0880 - Ethylamine solution

EMAIL THIS PAGE TO A FRIEND
09-0880 Sigma-Aldrich

Ethylamine solution

70 wt. % in H2O, CP

Synonym: Aminoethane, Monoethylamine

  • CAS Number 75-04-7

  • Linear Formula C2H5NH2

  • Molecular Weight 45.08

  •  Beilstein/REAXYS Number 505933

  •  MDL number MFCD00008160

  •  PubChem Substance ID 329748687

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Organic Bases, Synthetic Reagents More...
grade   CP
availability   available only in Japan
concentration   70 wt. % in H2O
density   0.81 g/mL at 20 °C
SMILES string   CCN
InChI   1S/C2H7N/c1-2-3/h2-3H2,1H3
InChI key   QUSNBJAOOMFDIB-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Respiratory system
RIDADR 
UN 2270 8(3) / PGII
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?