• USA Home
  • 12-1660 - Guanidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
12-1660 Sigma-Aldrich

Guanidine hydrochloride

SAJ special grade, ≥99.0%

Synonym: Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride

  • CAS Number 50-01-1

  • Linear Formula NH2C(=NH)NH2 · HCl

  • Molecular Weight 95.53

  •  Beilstein/REAXYS Number 3591990

  •  EC Number 200-002-3

  •  MDL number MFCD00013026

  •  PubChem Substance ID 329749647

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Cell Biology, Chaotropic Agents, Core Bioreagents,
grade   SAJ special grade
assay   ≥99.0%
availability   available only in Japan
mp   180-185 °C (lit.)
density   1.3 g/cm3 (lit.)
SMILES string   Cl[H].NC(N)=N
InChI   1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H
InChI key   PJJJBBJSCAKJQF-UHFFFAOYSA-N
Gene Information   human ... KCNA1(3736), KCNA10(3744), KCNA2(3737), KCNA3(3738), KCNA4(3739), KCNA5(3741), KCNA6(3742), KCNA7(3743), KCNB1(3745), KCNB2(9312), KCNC1(3746), KCNC2(3747), KCNC3(3748), KCNC4(3749), KCND1(3750), KCND2(3751), KCND3(3752), KCNF1(3754), KCNG1(3755), KCNG2(26251), KCNG3(170850), KCNG4(93107), KCNH1(3756), KCNH2(3757), KCNH3(23416), KCNH4(23415), KCNH5(27133), KCNH6(81033), KCNH7(90134), KCNH8(131096), KCNQ1(3784), KCNQ2(3785), KCNQ3(3786), KCNQ4(9132), KCNQ5(56479), KCNS1(3787), KCNS2(3788), KCNS3(3790), KCNV1(27012), KCNV2(169522)

Description

Application

Strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MF4300000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?