EMAIL THIS PAGE TO A FRIEND
15-1570 Sigma-Aldrich

Isopentyl acetate

SAJ first grade, ≥93.0%

Synonym: Acetic acid 3-methylbutyl ester, Isoamyl acetate

  • CAS Number 123-92-2

  • Linear Formula CH3COOCH2CH2CH(CH3)2

  • Molecular Weight 130.18

  •  Beilstein/REAXYS Number 1744750

  •  MDL number MFCD00008946

  •  PubChem Substance ID 329750756

Purchase

Properties

Related Categories SAJ First Grade (Japan only), Solvent by Application, Solvents, Solvents by Special Grades (Japan Customers Only)
grade   SAJ first grade
vapor density   4.5 (vs air)
vapor pressure   5 mmHg ( 25 °C)
InChI Key   MLFHJEHSLIIPHL-UHFFFAOYSA-N
assay   ≥93.0%
autoignition temp.   680 °F
expl. lim.   7.5 %
availability   available only in Japan
refractive index   n20/D 1.4 (lit.)
bp   142 °C/756 mmHg(lit.)
mp   −78 °C (lit.)
density   0.876 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Dispensette™ S
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Related Content

Pressure-Temperature Nomograph Interactive Tool

Use the interactive controls above to simplify calculations and improve the efficiency of your distillation or evaporation requirements.
Keywords: Evaporation, Vaporization

Properties of Solvents Table

The table below presents the physical characteristics of common solvents. Easily reference or compare solvent properties like boiling point, density, and refractive index all from one list.
Keywords: Gas chromatography, Green chemistry, High performance liquid chromatography, Melting, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?