• USA Home
  • 24-3765 - Poly(vinyl alcohol)

EMAIL THIS PAGE TO A FRIEND
24-3765 Sigma-Aldrich

Poly(vinyl alcohol)

SAJ first grade, degree of polymerization 1700

Purchase

Properties

Related Categories Cell Biology, Centrifugation Media, Core Bioreagents, Density Gradient Media and Cell Separation, Hematology and Histology,
grade   SAJ first grade
mol wt   degree of polymerization 1700
availability   available only in Japan
InChI   1S/C2H4O/c1-2-3/h2-3H,1H2
InChI key   IMROMDMJAWUWLK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
RTECS 
TR8100000
Flash Point(F) 
No data available
Flash Point(C) 
No data available

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?