EMAIL THIS PAGE TO A FRIEND
28-2130 Sigma-Aldrich

Sodium octanoate

SAJ first grade, ≥95.0%

Synonym: Caprylic acid sodium salt, Octanoic acid sodium salt, Sodium caprylate

  • CAS Number 1984-06-1

  • Linear Formula CH3(CH2)6COONa

  • Molecular Weight 166.19

  •  Beilstein/REAXYS Number 3597723

  •  MDL number MFCD00058511

  •  PubChem Substance ID 329753090

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Carbonyl Compounds, Carboxylic Acid Salts, Chemical Synthesis,
grade   SAJ first grade
assay   ≥95.0%
availability   available only in Japan
SMILES string   [Na+].CCCCCCCC([O-])=O
InChI   1S/C8H16O2.Na/c1-2-3-4-5-6-7-8(9)10;/h2-7H2,1H3,(H,9,10);/q;+1/p-1
InChI key   BYKRNSHANADUFY-UHFFFAOYSA-M
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?