EMAIL THIS PAGE TO A FRIEND
28-2760 Sigma-Aldrich

Sodium formate

JIS special grade, ≥98.0%

Synonym: Formic acid sodium salt

  • CAS Number 141-53-7

  • Linear Formula HCOONa

  • Molecular Weight 68.01

  •  Beilstein/REAXYS Number 3595134

  •  MDL number MFCD00013101

  •  PubChem Substance ID 329753116

Purchase

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Sodium, Synthetic Reagents
grade   JIS special grade
assay   ≥98.0%
availability   available only in Japan
mp   259-262 °C (lit.)
SMILES string   [Na+].[O-]C=O
InChI   1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1
InChI key   HLBBKKJFGFRGMU-UHFFFAOYSA-M
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?