EMAIL THIS PAGE TO A FRIEND
30-0320 Sigma-Aldrich

Terephthalic acid

SAJ special grade, ≥98.0%

Synonym: Benzene-1,4-dicarboxylic acid

  • CAS Number 100-21-0

  • Linear Formula C6H4-1,4-(CO2H)2

  • Molecular Weight 166.13

  •  Beilstein/REAXYS Number 1909333

  •  MDL number MFCD00002558

  •  PubChem Substance ID 329753411

Purchase

Properties

Related Categories Building Blocks, C8, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
grade   SAJ special grade
vapor pressure   <0.01 mmHg ( 20 °C)
InChI Key   KKEYFWRCBNTPAC-UHFFFAOYSA-N
assay   ≥98.0%
autoignition temp.   925 °F
availability   not available in Japan
mp   >300 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?