• USA Home
  • 30-0850 - Tetraethylenepentamine

EMAIL THIS PAGE TO A FRIEND
30-0850 Sigma-Aldrich

Tetraethylenepentamine

CP

Synonym: TEPA, Tetrene

  • CAS Number 112-57-2

  • Linear Formula (NH2CH2CH2NHCH2CH2)2NH

  • Molecular Weight 189.30

  •  Beilstein/REAXYS Number 506966

  •  MDL number MFCD00008168

  •  PubChem Substance ID 329753426

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks, Polyamines More...
grade   CP
vapor density   6.53 (vs air)
autoignition temp.   610 °F
availability   available only in Japan
refractive index   n20/D 1.505 (lit.)
mp   −40 °C (lit.)
density   0.998 g/mL at 25 °C (lit.)
SMILES string   NCCNCCNCCNCCN
InChI   1S/C8H23N5/c9-1-3-11-5-7-13-8-6-12-4-2-10/h11-13H,1-10H2
InChI key   FAGUFWYHJQFNRV-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
WGK Germany 
WGK 2
Flash Point(F) 
325.4 °F - closed cup
Flash Point(C) 
163 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?