• USA Home
  • 30-2460 - Tin(IV) chloride pentahydrate

EMAIL THIS PAGE TO A FRIEND
30-2460 Sigma-Aldrich

Tin(IV) chloride pentahydrate

SAJ special grade, ≥98.0%

Synonym: Tin tetrachloride

Purchase

Properties

grade   SAJ special grade
InChI Key   KHMOASUYFVRATF-UHFFFAOYSA-J
assay   ≥98.0%
availability   available only in Japan
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?