• USA Home
  • 30-3370 - p-Toluenesulfonic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
30-3370 Sigma-Aldrich

p-Toluenesulfonic acid monohydrate

CP

  • CAS Number 6192-52-5

  • Linear Formula CH3C6H4SO3H · H2O

  • Molecular Weight 190.22

  •  Beilstein Registry Number 3568023

  •  MDL number MFCD00142137

  •  PubChem Substance ID 329753475

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Organic Acids, Synthetic Reagents More...
grade   CP
vapor density   5.9 (vs air)
InChI Key   KJIFKLIQANRMOU-UHFFFAOYSA-N
availability   available only in Japan
mp   103-106 °C (lit.)
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?