EMAIL THIS PAGE TO A FRIEND
00380590 Supelco

(+)-Fenchone

primary reference standard

Synonym: (+)-1,3,3-Trimethyl-2-norbornanone, (1S)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one

  • CAS Number 4695-62-9

  • Empirical Formula (Hill Notation) C10H16O

  • Molecular Weight 152.23

  •  Beilstein Registry Number 2206555

  •  EC Number 225-160-0

  •  MDL number MFCD00070689

  •  PubChem Substance ID 329747056

Purchase

Properties

Related Categories A-F, A-L, All Alphabetically Sorted, Alphabetical Index of Phytochemical Standards, Analytical Standards,
grade   primary reference standard
InChI Key   LHXDLQBQYFFVNW-XCBNKYQSSA-N
CofA   certificate is enclosed in each package
shelf life   limited shelf life, expiry date on the label
bp   63-65 °C/13 mmHg(lit.)
mp   5-7 °C (lit.)
density   0.945 g/mL at 20 °C(lit.)

Description

Application

Reference Standard in the analysis of herbal medicinal products

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
RB7875200
Flash Point(F) 
150.8 °F
Flash Point(C) 
66 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?