EMAIL THIS PAGE TO A FRIEND
00760595 Sigma-Aldrich

Bilobalide

primary reference standard

  • CAS Number 33570-04-6

  • Empirical Formula (Hill Notation) C15H18O8

  • Molecular Weight 326.30

  •  MDL number MFCD00132880

  •  PubChem Substance ID 329747133

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Isoprenoids, Phyto Primary Reference Standards and Extracts (Secondary Reference Standards), Phytochemical Standards More...
Quality Level   100
grade   primary reference standard
CofA   certificate is enclosed in each package
shelf life   limited shelf life, expiry date on the label
storage temp.   −20°C
SMILES string   CC(C)(C)[C@]1(O)C[C@@H]2OC(=O)C[C@]23C(=O)O[C@@H]4OC(=O)[C@H](O)C134
InChI   1S/C15H18O8/c1-12(2,3)14(20)4-6-13(5-7(16)21-6)10(19)23-11-15(13,14)8(17)9(18)22-11/h6,8,11,17,20H,4-5H2,1-3H3/t6-,8-,11-,13+,14+,15?/m0/s1
InChI key   MOLPUWBMSBJXER-DXPZGIEZSA-N

Description

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
LV1665000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?