• USA Home
  • 00786 - Phenacyl 4-(bromomethyl)phenylacetate

EMAIL THIS PAGE TO A FRIEND
00786 Sigma-Aldrich

Phenacyl 4-(bromomethyl)phenylacetate

technical, ≥90% (GC)

Synonym: 4-(Bromomethyl)phenylacetic acid phenacyl ester

  • CAS Number 66270-97-1

  • Empirical Formula (Hill Notation) C17H15BrO3

  • Molecular Weight 347.20

  •  Beilstein/REAXYS Number 2292032

  •  MDL number MFCD00009632

  •  PubChem Substance ID 24844954

Purchase

Properties

grade   technical
assay   ≥90% (GC)
mp   84-87 °C (lit.)
  84-87 °C
SMILES string   BrCc1ccc(CC(=O)OCC(=O)c2ccccc2)cc1
InChI   1S/C17H15BrO3/c18-11-14-8-6-13(7-9-14)10-17(20)21-12-16(19)15-4-2-1-3-5-15/h1-9H,10-12H2
InChI key   BUQPIKAEKYNDAS-UHFFFAOYSA-N

Description

Other Notes

Building block for preparing amino acid PAM linker. The Pac ester is orthogonally stable to cleavage conditions of most protective groups used in FMOC-SPPS

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?