EMAIL THIS PAGE TO A FRIEND
01090595 Sigma-Aldrich

Hulupinic acid

primary reference standard

Synonym: 4,5-Dihydroxy-2,2-bis(3-methyl-2-butenyl)- 4-cyclopentene-1,3-dione

  • CAS Number 1891-42-5

  • Empirical Formula (Hill Notation) C15H20O4

  • Molecular Weight 264.32

  •  Beilstein/REAXYS Number 2124574

  •  MDL number MFCD16652587

  •  PubChem Substance ID 329747211

  •  NACRES NA.24

Purchase

Properties

Related Categories Aldehydes and Ketones, Alphabetical Index of Phytochemical Standards, Analytical Standards, Analytical/Chromatography, E - H,
grade   primary reference standard
InChI Key   KKNXLCGOZLVUHL-UHFFFAOYSA-N
CofA   certificate is enclosed in each package
shelf life   limited shelf life, expiry date on the label
Featured Industry   Food and Beverages

Description

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?