• USA Home
  • 03110 - N-Benzyl-N-ethylaniline

EMAIL THIS PAGE TO A FRIEND
03110 Sigma-Aldrich

N-Benzyl-N-ethylaniline

technical, ≥97.0% (NT)

Synonym: N-Ethyl-N-phenylbenzylamine

  • CAS Number 92-59-1

  • Linear Formula C6H5N(C2H5)CH2C6H5

  • Molecular Weight 211.30

  •  Beilstein Registry Number 2099852

  •  EC Number 202-169-8

  •  MDL number MFCD00009037

  •  PubChem Substance ID 329747556

Purchase

Properties

InChI Key   HSZCJVZRHXPCIA-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (NT)
refractive index   n20/D 1.595(lit.)
  n20/D 1.595
bp   163-164 °C/6 mmHg(lit.)
density   1.029 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2274 6.1 / PGIII
WGK Germany 
2
Flash Point(F) 
314.6 °F
Flash Point(C) 
157 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?