• USA Home
  • 03673 - Fmoc-β-Homoarg(Pmc)-OH

EMAIL THIS PAGE TO A FRIEND
03673 Sigma-Aldrich

Fmoc-β-Homoarg(Pmc)-OH

≥95.0% (HPLC)

Synonym: Nβ-Fmoc-Nω-(2,2,5,7,8-pentamethylchromane-6-sulfonyl)-L-β-homoarginine

  • CAS Number 700377-76-0

  • Empirical Formula (Hill Notation) C36H44N4O7S

  • Molecular Weight 676.82

  •  MDL number MFCD03788172

  •  PubChem Substance ID 329747762

  •  NACRES NA.26

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
Quality Level   100
assay   ≥95.0% (HPLC)
form   powder
application(s)   peptide synthesis: suitable
color   white to off-white
functional group   Fmoc
storage temp.   2-8°C
SMILES string   Cc1c(C)c(c(C)c2CCC(C)(C)Oc12)S(=O)(=O)NC(=N)NCCC[C@@H](CC(O)=O)NC(=O)OCC3c4ccccc4-c5ccccc35
InChI   1S/C36H44N4O7S/c1-21-22(2)33(23(3)25-16-17-36(4,5)47-32(21)25)48(44,45)40-34(37)38-18-10-11-24(19-31(41)42)39-35(43)46-20-30-28-14-8-6-12-26(28)27-13-7-9-15-29(27)30/h6-9,12-15,24,30H,10-11,16-20H2,1-5H3,(H,39,43)(H,41,42)(H3,37,38,40)/t24-/m0/s1
InChI key   MGKMEQWQKICWRD-DEOSSOPVSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?