• USA Home
  • 03841 - α-Cyano-4-hydroxycinnamic acid - α-Cyano-2,4-difluorocinnamic acid - α-Cyano-2,3,4,5,6-pentafluorocinnamic acid mixture

EMAIL THIS PAGE TO A FRIEND
03841 Supelco

α-Cyano-4-hydroxycinnamic acid - α-Cyano-2,4-difluorocinnamic acid - α-Cyano-2,3,4,5,6-pentafluorocinnamic acid mixture

matrix substance for MALDI-MS, ≥95.0% (sum of three components, HPLC)

Synonym: CHCA:DiFCCA:PentaFCCA

  •  NACRES NA.21

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Mass Spectrometry, Mass Spectrometry (MS), Matrices for MALDI Analysis,
Quality Level   100
grade   matrix substance for MALDI-MS
assay   ≥95.0% (sum of three components, HPLC)
analyte functional class(es)   drugs of abuse, ionic liquids (quantification)
analyte chemical class(es)   chlorinated lipids, lipids, peptides, phospholipids, phosphopeptides
application(s)   collision-induced dissociation MS/MS (CID-MS/MS): suitable
  matrix-enhanced secondary ion MS (ME-SIMS): suitable
solubility   methanol: 100 mg/10 mL, clear, colorless to light yellow

Description

Application

suitable as MALDI-Matrix material

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

900666 Bruker standard solvent, for MALDI MS, 50% ACN + 47.5% H2O + 2.5% TFA
P2613 ProteoMass P14R MALDI-MS Standard, vial of 10 nmol
C8857 ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
C8982 α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
A8346 ProteoMass ACTH Fragment 18-39 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 2,464.1989 Da
I6154 ProteoMass Insulin chain B oxidized MALDI-MS Standard, vial of 10 nmol
I6279 ProteoMass Insulin MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 5,729.6087 Da
A8471 ProteoMass Albumin MALDI-MS Standard, vial of 10 nmol, average mol wt 66,429.09 Da by calculation
A9096 ProteoMass Aldolase MALDI-MS Standard, vial of 10 nmol, average mol wt 39,211.28 Da by calculation
A8846 ProteoMass Angiotensin II MALDI-MS Standard, vial of 10 nmol
A8971 ProteoMass Apomyoglobin MALDI-MS Standard, vial of 10 nmol, (M+H+) 16,952.27 Da by calculation
B4181 ProteoMass Bradykinin Fragment 1-7 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 756.3997 Da
MSCAL1 ProteoMass Peptide and Protein MALDI-MS Calibration Kit, Peptide and Protein MALDI MS Calibration Standard
MSCAL2 ProteoMass Peptide MALDI-MS Calibration Kit, Peptide MALDI MS Calibration Standard
MSCAL3 ProteoMass Protein MALDI-MS Calibration Kit, Protein MALDI MS Calibration Standard
56197 3-Hydroxypicolinic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
54793 2-(4-Hydroxyphenylazo)benzoic acid, matrix substance for MALDI-MS, ≥99.5%
85429 Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
03565 MALDI validation set polystyrene Mp ~700 - 70 000, certified reference material
37468 2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5%
91928 2′,4′,6′-Trihydroxyacetophenone monohydrate, matrix substance for MALDI-MS, ≥99.5%
03597 MALDI validation set PS, PMMA, PDMS, PEG and PSS Mp ~4′300 - 6′500
87884 trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, matrix substance for MALDI-MS, ≥99.0% (HPLC)
60018 Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
29208 (N-Succinimidyloxycarbonylmethyl)tris(2,4,6-trimethoxyphenyl)phosphonium bromide, for protein sequence analysis (by MALDI-MS), ≥98.5%
46278 trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
85707 2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
70990 α-Cyano-4-hydroxycinnamic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
N3786 α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
39468 α-Cyano-4-hydroxycinnamic acid, matrix substance for MALDI-MS, Ultra pure
39319 2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
49508 Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
50862 Super-DHB, suitable for matrix substance for MALDI-MS, ≥99.0%
02241 2′,3′-Dideoxycytidine 5′-triphosphate sodium salt solution, for MALDI MS, ≥98.0% (HPLC)
MSCAL4 ProteoMass MALDI Calibration Kit for Exactive Series MS, LTQ XL and LTQ Hybrids, MALDI MS Calibration Standard
92817 9-Aminoacridine, matrix substance for MALDI-MS, ≥99.5% (HPLC)
56451 1,5-Diaminonaphthalene, matrix substance for MALDI-MS, ≥99.0% (HPLC)
50757 Micro-Array for Mass Spectrometry (MAMS), for single cell analysis
94141 4-Chloro-α-cyanocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
77646 α-Cyano-2,4-difluorocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
89063 4-Bromo-α-cyanocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
77081 α-Cyano-4-fluorocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
38419 α-Cyano-2,3,4,5,6-pentafluorocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
39379 4-Chloro-α-cyanocinnamic acid - α-Cyano-2,4-difluorocinnamic acid mixture, matrix substance for MALDI-MS, ≥95.0% (sum of both components, HPLC)
68914 4-Bromo-α-cyanocinnamic acid - 4-Chloro-α-cyanocinnamic acid mixture, matrix substance for MALDI-MS, ≥95.0% (sum of both components, HPLC)
05851 4-Aminoquinaldine, matrix substance for MALDI-MS, ≥99.0% (HPLC)
78246 Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
04973 Micro-Array for Mass Spectrometry (MAMS), for quantitative MALDI
94477 (E)-2-Cyano-3-(2-naphthyl)acrylic acid, matrix substance for MALDI-MS, ≥98.0% (HPLC)
83788 (2E)-3-(9-Anthryl)-2-cyanoacrylic acid, matrix substance for MALDI-MS, ≥97.0% (HPLC)
76884 Anthranilamide, matrix substance for MALDI-MS, ≥99.0% (HPLC)
56229 9-Nitroanthracene, matrix substance for MALDI-MS, ≥98.5% (HPLC)
PFS11 SpheriCal Mix, Peptide Low, for MALDI MS
PFS12 SpheriCal Mix, Peptide Medium, for MALDI MS
PFS13 SpheriCal Mix, Peptide High, for MALDI MS
PFS14 SpheriCal Mix, Protein Low, for MALDI MS
PFS15 SpheriCal Mix, Protein Medium, for MALDI MS
PFS31 SpheriCal Aqua, Peptide Medium, for MALDI MS
PFS32 SpheriCal Aqua, Peptide High, for MALDI MS
PFS33 SpheriCal Aqua, Protein Low, for MALDI MS
PFS20 SpheriCal Neat, Peptide Low, for MALDI MS
PFS21 SpheriCal Neat, Peptide Medium, for MALDI MS
PFS22 SpheriCal Neat, Peptide High, for MALDI MS
PFS23 SpheriCal Neat, Protein Low, for MALDI MS
PFS24 SpheriCal Neat, Protein Medium, for MALDI MS
PFS30 SpheriCal Aqua, Peptide Low, for MALDI MS
69028 4-Phenyl-α-cyanocinnamamide, matrix substance for MALDI-MS, ≥98.5% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?