EMAIL THIS PAGE TO A FRIEND
03979 Sigma-Aldrich

Weigert’s solution

for microscopy

Synonym: Fuchsin solution

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Microscopy Reagents, Stains and Dyes
grade   for microscopy
InChI Key   DVSQBUUDEVNABC-OULXEKPRSA-N
impurities   diamond fuchsin
  ethanol
density   0.82 g/mL at 20 °C

Description

Application

for coloring of elastic fibres

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?