• USA Home
  • 05169 - N-Methyl-2-pyrrolecarboxaldehyde

EMAIL THIS PAGE TO A FRIEND
05169 Supelco

N-Methyl-2-pyrrolecarboxaldehyde

analytical standard

  • CAS Number 1192-58-1

  • Empirical Formula (Hill Notation) C6H7NO

  • Molecular Weight 109.13

  •  Beilstein/REAXYS Number 107811

  •  EC Number 214-755-0

  •  MDL number MFCD00003087

  •  PubChem Substance ID 329747651

  •  NACRES NA.24

Purchase

Properties

Related Categories Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Analytical Standards, Analytical/Chromatography, Coffee,
grade   analytical standard
InChI Key   OUKQTRFCDKSEPL-UHFFFAOYSA-N
assay   ≥98.0% (GC)
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.560-1.564
bp   87-90 °C/22 mmHg(lit.)
density   1.016 g/mL at 25 °C (lit.)
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
UX9350000
Flash Point(F) 
161.6 °F
Flash Point(C) 
72 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?