• USA Home
  • 05170 - L-Alanine ethyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
05170 Sigma-Aldrich

L-Alanine ethyl ester hydrochloride

puriss., ≥99.0% (TLC)

  • CAS Number 1115-59-9

  • Linear Formula CH3CH(NH2)COOC2H5 · HCl

  • Molecular Weight 153.61

  •  Beilstein Registry Number 3594395

  •  EC Number 214-225-9

  •  MDL number MFCD00063662

  •  PubChem Substance ID 329747652

Purchase

Properties

Related Categories Alanine, Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   JCXLZWMDXJFOOI-WCCKRBBISA-N
grade   puriss.
assay   ≥99.0% (TLC)
optical activity   [α]20/D +2.9±0.3°, c = 2.5% in H2O
impurities   ~1% water
mp   78-80 °C (dec.)(lit.)
  ~80 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

855669 L-Alanine ethyl ester hydrochloride, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?